Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173907345-173907612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389405 c.1165G>C p.E389Q Chr.1: 173907503-173907503 on Build GRCh38
4843553 c.1172G>A p.R391Q Chr.1: 173907496-173907496 on Build GRCh38
3478547 c.1201C>A p.H401N Chr.1: 173907467-173907467 on Build GRCh38
1225294 c.1178A>C p.D393A Chr.1: 173907490-173907490 on Build GRCh38
4025224 c.1214T>G p.L405R Chr.1: 173907454-173907454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751704392 Chr.1: 173907552-173907552 on Build GRCh38
rs372611817 Chr.1: 173907474-173907474 on Build GRCh38
rs375346550 Chr.1: 173907519-173907519 on Build GRCh38
rs757611426 Chr.1: 173907447-173907447 on Build GRCh38
rs367854110 Chr.1: 173907405-173907405 on Build GRCh38
rs761918672 Chr.1: 173907560-173907560 on Build GRCh38
rs920003404 Chr.1: 173907462-173907462 on Build GRCh38
rs190912722 Chr.1: 173907575-173907575 on Build GRCh38
rs550785920 Chr.1: 173907419-173907419 on Build GRCh38
rs771854239 Chr.1: 173907542-173907542 on Build GRCh38
rs759275331 Chr.1: 173907504-173907504 on Build GRCh38
rs560610974 Chr.1: 173907509-173907509 on Build GRCh38
rs483352858 Chr.1: 173907521-173907521 on Build GRCh38
rs763052075 Chr.1: 173907529-173907529 on Build GRCh38
rs749848792 Chr.1: 173907475-173907475 on Build GRCh38
rs764115074 Chr.1: 173907549-173907549 on Build GRCh38
rs770580443 Chr.1: 173907500-173907500 on Build GRCh38
rs201541724 Chr.1: 173907496-173907496 on Build GRCh38
rs483352859 Chr.1: 173907455-173907455 on Build GRCh38
rs762965029 Chr.1: 173907424-173907424 on Build GRCh38
rs677 Chr.1: 173907423-173907423 on Build GRCh38
rs368902256 Chr.1: 173907450-173907450 on Build GRCh38
rs878854018 Chr.1: 173907514-173907514 on Build GRCh38
rs551665576 Chr.1: 173907578-173907578 on Build GRCh38
rs777327592 Chr.1: 173907484-173907484 on Build GRCh38
rs1004108836 Chr.1: 173907541-173907541 on Build GRCh38
rs899914657 Chr.1: 173907443-173907443 on Build GRCh38
rs2759328 Chr.1: 173907567-173907567 on Build GRCh38
rs765121710 Chr.1: 173907412-173907412 on Build GRCh38
rs541549098 Chr.1: 173907583-173907583 on Build GRCh38
rs746504812 Chr.1: 173907489-173907489 on Build GRCh38
rs776344892 Chr.1: 173907402-173907402 on Build GRCh38
rs540730281 Chr.1: 173907452-173907452 on Build GRCh38
rs764313616 Chr.1: 173907431-173907431 on Build GRCh38
rs756459428 Chr.1: 173907461-173907461 on Build GRCh38
rs542881762 Chr.1: 173907528-173907528 on Build GRCh38
rs375038144 Chr.1: 173907434-173907434 on Build GRCh38
rs776277707 Chr.1: 173907501-173907501 on Build GRCh38
rs769120458 Chr.1: 173907483-173907483 on Build GRCh38

View Full Product Details