Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014349.2 | NP_055164.1 | 3 - 4 | 713 | 117 |
NM_030644.1 | NP_085147.1 | 4 - 5 | 597 | 117 | |
NM_145639.1 | NP_663614.1 | 3 - 4 | 484 | 117 | |
NM_145640.2 | NP_663615.1 | 2 - 3 | 387 | 117 | |
NM_145641.2 | NP_663616.1 | 4 - 5 | 1896 | 117 | |
NM_145642.2 | NP_663617.1 | 3 - 4 | 1833 | 117 | |
XM_006724324.1 | XP_006724387.1 | 4 - 5 | 475 | 117 | |
XM_006724325.2 | XP_006724388.1 | 3 - 4 | 386 | 117 | |
XM_017028946.1 | XP_016884435.1 | 3 - 4 | 313 | 117 | |
XM_017028947.1 | XP_016884436.1 | 4 - 5 | 426 | 117 | |
XM_017028948.1 | XP_016884437.1 | 5 - 6 | 730 | 117 | |
XM_017028949.1 | XP_016884438.1 | 5 - 6 | 615 | 117 | |
XM_017028950.1 | XP_016884439.1 | 4 - 5 | 1320 | 117 | |
XM_017028951.1 | XP_016884440.1 | 3 - 4 | 1207 | 117 | |
XM_017028952.1 | XP_016884441.1 | 2 - 3 | 233 | 117 | |
GenBank mRNA | AF070675.1 | - | 3 - 4 | 553 | 117 |
AF305227.1 | - | 3 - 4 | 340 | 117 | |
AK025349.1 | - | 3 - 4 | 310 | 117 | |
AY014902.1 | - | 4 - 5 | 597 | 117 | |
AY014903.1 | - | 3 - 4 | 484 | 117 | |
AY014904.1 | - | 2 - 3 | 387 | 117 | |
AY014905.1 | - | 3 - 4 | 713 | 117 | |
AY014906.1 | - | 4 - 5 | 1896 | 117 | |
AY014907.1 | - | 3 - 4 | 1833 | 117 | |
BC042918.1 | - | 3 - 4 | 289 | 117 | |
BC050596.1 | - | 3 - 4 | 344 | 117 | |
BF797347.1 | - | 2 - 3 | 248 | 117 | |
CR456379.1 | - | 2 - 3 | 207 | 117 | |
DA878288.1 | - | 2 - 3 | 317 | 117 |