Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207067.1 | NP_001193996.1 | 5 - 6 | 862 | 150 |
NM_001207069.1 | NP_001193998.1 | 5 - 6 | 542 | 150 | |
NM_001321688.1 | NP_001308617.1 | 5 - 6 | 442 | 150 | |
NM_001321690.1 | NP_001308619.1 | 5 - 6 | 556 | 150 | |
NM_001321691.1 | NP_001308620.1 | 5 - 6 | 486 | 150 | |
NM_001321693.1 | NP_001308622.1 | 4 - 5 | 309 | 150 | |
NM_001321694.1 | NP_001308623.1 | 5 - 6 | 417 | 150 | |
NM_014670.3 | NP_055485.2 | 5 - 6 | 862 | 150 | |
XM_017005393.1 | XP_016860882.1 | 2 - 3 | 155 | 150 | |
GenBank mRNA | AK096489.1 | - | 4 - 5 | 417 | 150 |
AK223245.1 | - | 5 - 6 | 471 | 150 | |
AK297227.1 | - | 5 - 6 | 555 | 150 | |
AK301511.1 | - | 5 - 6 | 542 | 150 | |
AL833518.1 | - | 5 - 6 | 818 | 150 | |
BC001804.1 | - | 5 - 6 | 484 | 150 | |
BC026303.1 | - | 5 - 6 | 150 | ||
D13630.1 | - | 5 - 6 | 487 | 150 |