Assay Design



Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
84321
Gene Name
THO complex 3
Gene Aliases
THO3, hTREX45
Chromosome Location
Chr.5: 175959531 - 176034905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032361.3 NP_115737.1 1 - 2 376 65
GenBank mRNA BC006849.1 - 1 - 2 338 65
BC066325.1 - 1 - 2 333 65
BC068499.1 - 1 - 2 295 65
BG502664.1 - 1 - 2 97 65
HY009859.1 - 1 - 2 377 65

Gene Symbol
LOC728554
Entrez Gene ID
728554
Gene Name
THO complex 3 pseudogene
Gene Aliases
-
Chromosome Location
Chr.5: 177875261 - 177884268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003615.2 1 - 2 602 65

View Full Product Details