Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000972.2 | NP_000963.1 | 2 - 3 | 155 | 71 |
GenBank mRNA | AK291123.1 | - | 2 - 3 | 147 | 71 |
AK311743.1 | - | 2 - 3 | 151 | 71 | |
BC005128.1 | - | 1 - 1 | 138 | 71 | |
BC021052.1 | - | 1 - 1 | 129 | 71 | |
BC021979.2 | - | 1 - 1 | 127 | 71 | |
BC023594.2 | - | 2 - 3 | 186 | 71 | |
BC023624.2 | - | 2 - 3 | 191 | 71 | |
BC032533.1 | - | 1 - 1 | 174 | 71 | |
BC071900.1 | - | 1 - 1 | 127 | 71 | |
BC071901.1 | - | 1 - 2 | 135 | 71 | |
BC073802.1 | - | 1 - 1 | 126 | 71 | |
BC105290.1 | - | 1 - 1 | 152 | 71 | |
M36072.1 | - | 2 - 3 | 156 | 71 | |
X06705.1 | - | 2 - 3 | 155 | 71 |