Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31064134-31064389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
33194 c.275G>T p.G92V Chr.8: 31064354-31064354 on Build GRCh38
750649 c.254G>T p.W85L Chr.8: 31064333-31064333 on Build GRCh38
223243 c.223G>A p.D75N Chr.8: 31064302-31064302 on Build GRCh38
6113022 c.264A>T p.L88F Chr.8: 31064343-31064343 on Build GRCh38
2958606 c.252G>A p.E84E Chr.8: 31064331-31064331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780286946 Chr.8: 31064238-31064238 on Build GRCh38
rs6983980 Chr.8: 31064236-31064236 on Build GRCh38
rs551533634 Chr.8: 31064181-31064181 on Build GRCh38
rs770781219 Chr.8: 31064271-31064271 on Build GRCh38
rs747587698 Chr.8: 31064304-31064305 on Build GRCh38
rs1042585683 Chr.8: 31064186-31064186 on Build GRCh38
rs566489064 Chr.8: 31064195-31064195 on Build GRCh38
rs773717744 Chr.8: 31064338-31064338 on Build GRCh38
rs760452488 Chr.8: 31064320-31064320 on Build GRCh38
rs879170656 Chr.8: 31064235-31064235 on Build GRCh38
rs148758902 Chr.8: 31064349-31064349 on Build GRCh38
rs566693857 Chr.8: 31064298-31064298 on Build GRCh38
rs368380436 Chr.8: 31064272-31064272 on Build GRCh38
rs773841160 Chr.8: 31064244-31064245 on Build GRCh38
rs758988621 Chr.8: 31064288-31064288 on Build GRCh38
rs754221635 Chr.8: 31064355-31064355 on Build GRCh38
rs918632815 Chr.8: 31064204-31064204 on Build GRCh38
rs3087407 Chr.8: 31064254-31064254 on Build GRCh38
rs374160869 Chr.8: 31064256-31064256 on Build GRCh38
rs772594712 Chr.8: 31064330-31064344 on Build GRCh38
rs536011576 Chr.8: 31064257-31064257 on Build GRCh38
rs7003607 Chr.8: 31064237-31064237 on Build GRCh38
rs777837851 Chr.8: 31064216-31064216 on Build GRCh38
rs878854135 Chr.8: 31064300-31064300 on Build GRCh38
rs1023919016 Chr.8: 31064264-31064264 on Build GRCh38
rs144670883 Chr.8: 31064308-31064308 on Build GRCh38
rs749107393 Chr.8: 31064270-31064270 on Build GRCh38
rs11574197 Chr.8: 31064199-31064199 on Build GRCh38
rs758351156 Chr.8: 31064176-31064176 on Build GRCh38

View Full Product Details