Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025105.2 | NP_001020276.1 | 1 - 2 | 717 | 72 |
NM_001271741.1 | NP_001258670.1 | 1 - 2 | 717 | 72 | |
NM_001892.5 | NP_001883.4 | 1 - 2 | 717 | 72 | |
GenBank mRNA | AF218004.1 | - | 1 - 2 | 515 | 72 |
AK294942.1 | - | 2 - 3 | 304 | 72 | |
AK303794.1 | - | 2 - 3 | 350 | 72 | |
BC007246.1 | - | 1 - 2 | 538 | 72 | |
BC008717.2 | - | 1 - 2 | 259 | 72 | |
BC021971.2 | - | 1 - 2 | 536 | 72 | |
BC025371.2 | - | 1 - 2 | 536 | 72 | |
BT019829.1 | - | 1 - 2 | 129 | 72 | |
BT019830.1 | - | 1 - 2 | 129 | 72 | |
CR542206.1 | - | 1 - 2 | 129 | 72 | |
DQ082865.1 | - | 1 - 2 | 129 | 72 | |
EU668329.1 | - | 1 - 2 | 129 | 72 | |
L37042.1 | - | 1 - 2 | 340 | 72 |