Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033449.2 | NP_258260.1 | 13 - 14 | 1368 | 69 |
XM_005268524.4 | XP_005268581.1 | 13 - 14 | 1395 | 69 | |
XM_006714803.3 | XP_006714866.1 | 12 - 13 | 1272 | 69 | |
XM_011537698.2 | XP_011536000.1 | 13 - 14 | 1401 | 69 | |
XM_011537700.2 | XP_011536002.1 | 13 - 14 | 1402 | 69 | |
XM_017010013.1 | XP_016865502.1 | 13 - 14 | 1401 | 69 | |
GenBank mRNA | AK000007.1 | - | 13 - 14 | 1352 | 69 |
AY217346.1 | - | 13 - 14 | 1368 | 69 | |
AY358478.1 | - | 4 - 5 | 986 | 69 | |
BC047016.1 | - | 12 - 13 | 1295 | 69 |