Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040134.1 | NP_001035224.1 | 1 - 2 | 200 | 60 |
NM_002579.2 | NP_002570.2 | 1 - 2 | 200 | 60 | |
GenBank mRNA | AK299809.1 | - | 1 - 2 | 56 | 60 |
AK315168.1 | - | 1 - 2 | 52 | 60 | |
BC032449.1 | - | 1 - 2 | 48 | 60 | |
BU508077.1 | - | - | 200 | 60 | |
Y14770.1 | - | 1 - 2 | 33 | 60 |