Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004304.4 | NP_004295.2 | 2 - 3 | 1743 | 106 |
GenBank mRNA | AB209477.4 | - | 2 - 3 | 1280 | 106 |
EU788003.1 | - | 2 - 3 | 1698 | 106 | |
EU788004.1 | - | 2 - 3 | 1698 | 106 | |
EU788005.1 | - | 2 - 3 | 1698 | 106 | |
EU788006.1 | - | 2 - 3 | 1698 | 106 | |
EU788007.1 | - | 2 - 3 | 1698 | 106 | |
EU788008.1 | - | 2 - 3 | 1698 | 106 | |
EU788009.1 | - | 2 - 3 | 1698 | 106 | |
EU788010.1 | - | 2 - 3 | 1698 | 106 | |
FJ838786.1 | - | 2 - 3 | 1698 | 106 | |
FJ838787.1 | - | 2 - 3 | 1698 | 106 | |
GU128152.1 | - | 2 - 3 | 791 | 106 | |
GU128153.1 | - | 2 - 3 | 791 | 106 | |
GU128154.1 | - | 2 - 3 | 791 | 106 | |
GU128155.1 | - | 2 - 3 | 791 | 106 | |
GU128156.1 | - | 2 - 3 | 791 | 106 | |
GU128157.1 | - | 2 - 3 | 791 | 106 | |
U62540.1 | - | 2 - 3 | 1702 | 106 | |
U66559.1 | - | 2 - 3 | 939 | 106 |