Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006147.3 | NP_006138.1 | 3 - 4 | 484 | 75 |
GenBank mRNA | AF027292.1 | - | 2 - 3 | 248 | 75 |
AK312857.1 | - | 3 - 4 | 384 | 75 | |
BC014852.1 | - | 3 - 4 | 353 | 75 | |
BI770148.1 | - | 3 - 4 | 443 | 75 | |
DA016081.1 | - | 3 - 4 | 484 | 75 |