Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256820.1 | NP_001243749.1 | 4 - 5 | 484 | 64 |
NM_001256821.1 | NP_001243750.1 | 5 - 6 | 511 | 64 | |
NM_006912.5 | NP_008843.1 | 5 - 6 | 633 | 64 | |
GenBank mRNA | AF084462.1 | - | 5 - 6 | 544 | 64 |
AF493923.1 | - | 4 - 5 | 430 | 64 | |
AK298768.1 | - | 4 - 5 | 484 | 64 | |
AK314239.1 | - | 5 - 6 | 609 | 64 | |
BC104186.2 | - | 5 - 6 | 504 | 64 | |
BC104187.2 | - | 5 - 6 | 504 | 64 | |
BI916790.1 | - | 5 - 6 | 494 | 64 | |
BM546985.1 | - | 5 - 6 | 627 | 64 | |
BQ918446.1 | - | 5 - 6 | 511 | 64 | |
CR407639.1 | - | 4 - 5 | 430 | 64 | |
U71203.1 | - | 5 - 6 | 565 | 64 | |
U78165.1 | - | 5 - 6 | 565 | 64 | |
Y07566.1 | - | 5 - 6 | 575 | 64 |