Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012454.3 | NP_036586.2 | 9 - 10 | 2733 | 86 |
GenBank mRNA | AF120323.1 | - | 5 - 6 | 703 | 86 |
AK123859.1 | - | 10 - 11 | 2828 | 86 | |
AK125459.1 | - | 6 - 7 | 1192 | 86 | |
AK302860.1 | - | 5 - 6 | 644 | 86 | |
AL122086.1 | - | 4 - 5 | 562 | 86 | |
BC137275.1 | - | 8 - 9 | 2606 | 86 |