Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017882.2 | NP_060352.1 | 2 - 3 | 358 | 104 |
GenBank mRNA | AK000568.1 | - | 2 - 3 | 303 | 104 |
AK027604.1 | - | 2 - 3 | 326 | 104 | |
AK124013.1 | - | 2 - 3 | 632 | 104 | |
AK291175.1 | - | 2 - 3 | 285 | 104 | |
AK293155.1 | - | 2 - 3 | 286 | 104 | |
AK293197.1 | - | 2 - 3 | 534 | 104 | |
BC010849.1 | - | 2 - 3 | 272 | 104 | |
BC013130.1 | - | 2 - 3 | 209 | 104 | |
CR457244.1 | - | 2 - 3 | 200 | 104 | |
KU178671.1 | - | 2 - 3 | 200 | 104 | |
KU178672.1 | - | 2 - 3 | 200 | 104 | |
KU178674.1 | - | 2 - 3 | 200 | 104 |