Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81706184-81706433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3030136 c.1711G>A p.V571I Chr.7: 81706333-81706333 on Build GRCh38
4529834 c.1646G>A p.G549E Chr.7: 81706398-81706398 on Build GRCh38
3431781 c.1617-2A>T p.? Chr.7: 81706429-81706429 on Build GRCh38
4529927 c.1654G>A p.D552N Chr.7: 81706390-81706390 on Build GRCh38
6937190 c.1676A>G p.E559G Chr.7: 81706368-81706368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144086354 Chr.7: 81706370-81706370 on Build GRCh38
rs752743857 Chr.7: 81706346-81706346 on Build GRCh38
rs779933934 Chr.7: 81706381-81706381 on Build GRCh38
rs778704152 Chr.7: 81706369-81706369 on Build GRCh38
rs763167149 Chr.7: 81706319-81706319 on Build GRCh38
rs1052238811 Chr.7: 81706229-81706229 on Build GRCh38
rs773824390 Chr.7: 81706313-81706313 on Build GRCh38
rs201395847 Chr.7: 81706298-81706298 on Build GRCh38
rs752861058 Chr.7: 81706392-81706392 on Build GRCh38
rs370757602 Chr.7: 81706372-81706372 on Build GRCh38
rs376043671 Chr.7: 81706357-81706357 on Build GRCh38
rs946475322 Chr.7: 81706307-81706307 on Build GRCh38
rs759776706 Chr.7: 81706336-81706336 on Build GRCh38
rs559990458 Chr.7: 81706359-81706359 on Build GRCh38
rs766769838 Chr.7: 81706329-81706329 on Build GRCh38
rs576420401 Chr.7: 81706243-81706243 on Build GRCh38
rs935150791 Chr.7: 81706224-81706224 on Build GRCh38
rs765831517 Chr.7: 81706299-81706299 on Build GRCh38
rs138548003 Chr.7: 81706340-81706340 on Build GRCh38
rs192078086 Chr.7: 81706395-81706395 on Build GRCh38
rs148098479 Chr.7: 81706382-81706382 on Build GRCh38
rs373049064 Chr.7: 81706281-81706281 on Build GRCh38
rs768245636 Chr.7: 81706250-81706250 on Build GRCh38
rs751689885 Chr.7: 81706331-81706331 on Build GRCh38
rs893517246 Chr.7: 81706279-81706279 on Build GRCh38
rs527424302 Chr.7: 81706220-81706220 on Build GRCh38

View Full Product Details