Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000302.3 | NP_000293.2 | 13 - 14 | 1579 | 62 |
NM_001316320.1 | NP_001303249.1 | 14 - 15 | 1720 | 62 | |
GenBank mRNA | AK294688.1 | - | 13 - 14 | 1410 | 62 |
AK299150.1 | - | 14 - 15 | 1653 | 62 | |
AK312243.1 | - | 13 - 14 | 1512 | 62 | |
AK316285.1 | - | 14 - 15 | 1656 | 62 | |
BC016657.2 | - | 13 - 14 | 1483 | 62 | |
M98252.1 | - | 13 - 14 | 1490 | 62 |