Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000673.4 | NP_000664.2 | 5 - 6 | 844 | 57 |
NM_001166504.1 | NP_001159976.1 | 5 - 6 | 657 | 57 | |
GenBank mRNA | AK223472.1 | - | 5 - 6 | 638 | 57 |
AK301657.1 | - | 5 - 6 | 620 | 57 | |
AK301696.1 | - | 5 - 6 | 657 | 57 | |
AK310811.1 | - | 4 - 5 | 608 | 57 | |
AK312854.1 | - | 5 - 6 | 767 | 57 | |
BC131512.1 | - | 5 - 6 | 639 | 57 | |
CD014081.1 | - | 6 - 7 | 720 | 57 | |
L33179.1 | - | 5 - 6 | 567 | 57 | |
U07821.1 | - | 5 - 6 | 651 | 57 | |
U09623.1 | - | 5 - 6 | 585 | 57 | |
X76342.1 | - | 5 - 6 | 627 | 57 |