Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033521.1 | NP_001028693.1 | 2 - 3 | 373 | 94 |
NM_001033522.1 | NP_001028694.1 | 2 - 3 | 370 | 94 | |
NM_001324.2 | NP_001315.1 | 2 - 3 | 401 | 94 | |
XM_011528600.1 | XP_011526902.1 | 2 - 3 | 389 | 94 | |
GenBank mRNA | AA037145.1 | - | 2 - 3 | 402 | 94 |
AK293182.1 | - | 3 - 4 | 235 | 94 | |
AK312774.1 | - | 2 - 3 | 274 | 94 | |
BC001011.2 | - | 2 - 3 | 262 | 94 | |
BC007425.2 | - | 2 - 3 | 262 | 94 | |
BI547689.1 | - | 2 - 3 | 378 | 94 | |
BT007138.1 | - | 1 - 2 | 173 | 94 | |
BU189971.1 | - | 2 - 3 | 367 | 94 | |
BU934383.1 | - | - | 368 | 94 | |
CF272312.1 | - | 2 - 3 | 370 | 94 | |
L02547.1 | - | 2 - 3 | 354 | 94 |