Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002532.5 | NP_002523.2 | 11 - 12 | 1729 | 144 |
XM_017024694.1 | XP_016880183.1 | 12 - 13 | 1779 | 144 | |
GenBank mRNA | AK225247.1 | - | 11 - 12 | 1649 | 144 |
AK308142.1 | - | 11 - 12 | 1541 | 144 | |
BC000335.2 | - | 11 - 12 | 1642 | 144 | |
Y08612.2 | - | 11 - 12 | 1688 | 144 |