Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320653.1 | NP_001307582.1 | 7 - 8 | 1276 | 97 |
NM_002532.5 | NP_002523.2 | 7 - 8 | 1276 | 97 | |
XM_011523893.2 | XP_011522195.1 | 8 - 9 | 1326 | 97 | |
XM_017024694.1 | XP_016880183.1 | 8 - 9 | 1326 | 97 | |
GenBank mRNA | AK225247.1 | - | 7 - 8 | 1196 | 97 |
AK298158.1 | - | 8 - 9 | 1053 | 97 | |
AK299000.1 | - | 7 - 8 | 1199 | 97 | |
AK308142.1 | - | 7 - 8 | 1088 | 97 | |
BC000335.2 | - | 7 - 8 | 1189 | 97 | |
Y08612.2 | - | 7 - 8 | 1235 | 97 |