Assay Design
Gene Transcripts
- Gene Symbol
- T
- Entrez Gene ID
-
6862
- Gene Name
- T brachyury transcription factor
- Gene Aliases
-
SAVA, TFT
- Chromosome Location
-
Chr.6: 166157644 - 166168669 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003181.3 |
NP_003172.1 |
6 - 7 |
1249 |
60 |
GenBank mRNA |
AJ001699.1 |
- |
5 - 6 |
889 |
60 |
View Full Product Details