Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144761.1 | NP_001138233.1 | 5 - 6 | 352 | 58 |
NM_001144762.1 | NP_001138234.1 | 4 - 5 | 270 | 58 | |
NM_001300846.1 | NP_001287775.1 | 4 - 5 | 526 | 58 | |
NM_003260.4 | NP_003251.2 | 4 - 5 | 492 | 58 | |
XM_006722864.2 | XP_006722927.1 | 4 - 5 | 434 | 58 | |
XM_011528230.1 | XP_011526532.1 | 4 - 5 | 434 | 58 | |
GenBank mRNA | AK293407.1 | - | 4 - 5 | 272 | 58 |
AK303102.1 | - | 4 - 5 | 368 | 58 | |
AK308137.1 | - | 5 - 6 | 352 | 58 | |
AK308608.1 | - | 4 - 5 | 244 | 58 | |
AK312285.1 | - | 4 - 5 | 338 | 58 | |
BC017364.2 | - | 4 - 5 | 349 | 58 | |
M99436.1 | - | 4 - 5 | 254 | 58 |