Assay Design
Gene Transcripts
- Gene Symbol
- KALRN
- Entrez Gene ID
-
8997
- Gene Name
- kalirin, RhoGEF kinase
- Gene Aliases
-
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
- Chromosome Location
-
Chr.3: 124033341 - 124726325 on Build GRCh38
View Full Product Details