Assay Design



Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 23 - 24 4004 73
NM_001322988.1 NP_001309917.1 23 - 24 4004 73
NM_001322989.1 NP_001309918.1 23 - 24 3977 73
NM_001322990.1 NP_001309919.1 23 - 24 3965 73
NM_001322991.1 NP_001309920.1 23 - 24 3938 73
NM_001322992.1 NP_001309921.1 22 - 23 3854 73
NM_003947.5 NP_003938.1 23 - 24 4004 73
XM_006713810.3 XP_006713873.1 23 - 24 4249 73
XM_006713811.3 XP_006713874.1 23 - 24 4222 73
XM_006713812.3 XP_006713875.1 23 - 24 4210 73
XM_006713813.3 XP_006713876.1 23 - 24 4183 73
XM_006713814.3 XP_006713877.1 23 - 24 4249 73
XM_006713815.3 XP_006713878.1 23 - 24 4252 73
XM_011513279.2 XP_011511581.1 23 - 24 4249 73
XM_011513280.2 XP_011511582.1 23 - 24 4250 73
XM_011513281.2 XP_011511583.1 23 - 24 26871 73
XM_011513283.2 XP_011511585.1 23 - 24 4246 73
XM_011513285.2 XP_011511587.1 23 - 24 4245 73
XM_017007429.1 XP_016862918.1 23 - 24 4249 73
XM_017007430.1 XP_016862919.1 23 - 24 4249 73
XM_017007431.1 XP_016862920.1 23 - 24 4245 73
XM_017007432.1 XP_016862921.1 23 - 24 4245 73
XM_017007433.1 XP_016862922.1 23 - 24 4225 73
XM_017007434.1 XP_016862923.1 23 - 24 4213 73
XM_017007435.1 XP_016862924.1 23 - 24 4186 73
GenBank mRNA AK131379.1 - 13 - 14 2065 73
AK314126.1 - 23 - 24 3977 73
U94190.1 - 23 - 24 3947 73

View Full Product Details