Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 11 - 12 | 2081 | 73 |
XM_005270276.4 | XP_005270333.1 | 11 - 12 | 2035 | 73 | |
XM_006718056.3 | XP_006718119.1 | 11 - 12 | 2044 | 73 | |
XM_006718057.3 | XP_006718120.1 | 11 - 12 | 2051 | 73 | |
XM_011540341.2 | XP_011538643.1 | 11 - 12 | 2205 | 73 | |
XM_011540342.1 | XP_011538644.1 | 11 - 12 | 1783 | 73 | |
XM_011540344.2 | XP_011538646.1 | 10 - 11 | 1690 | 73 | |
XM_011540345.1 | XP_011538647.1 | 10 - 11 | 1815 | 73 | |
XM_011540346.2 | XP_011538648.1 | 11 - 12 | 2050 | 73 | |
XM_011540347.2 | XP_011538649.1 | 7 - 8 | 1261 | 73 | |
XM_017016913.1 | XP_016872402.1 | 11 - 12 | 1744 | 73 | |
XM_017016914.1 | XP_016872403.1 | 11 - 12 | 2074 | 73 | |
XM_017016915.1 | XP_016872404.1 | 7 - 8 | 1945 | 73 | |
GenBank mRNA | AB011155.2 | - | 11 - 12 | 2111 | 73 |
AB091676.1 | - | 9 - 10 | 1546 | 73 | |
AK055172.1 | - | 7 - 8 | 999 | 73 | |
AK122931.1 | - | 12 - 13 | 2013 | 73 | |
BC117695.2 | - | 3 - 4 | 348 | 73 | |
BC146794.1 | - | 11 - 12 | 2111 | 73 | |
BX649131.1 | - | 7 - 8 | 1208 | 73 |