Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165946.1 | NP_001159418.1 | 8 - 9 | 868 | 70 |
NM_001322793.1 | NP_001309722.1 | 18 - 19 | 2284 | 70 | |
NM_001322794.1 | NP_001309723.1 | 18 - 19 | 2167 | 70 | |
NM_001322795.1 | NP_001309724.1 | 19 - 20 | 2466 | 70 | |
NM_001322796.1 | NP_001309725.1 | 18 - 19 | 2448 | 70 | |
NM_001322797.1 | NP_001309726.1 | 8 - 9 | 868 | 70 | |
NM_004969.3 | NP_004960.2 | 18 - 19 | 2284 | 70 | |
NR_136399.1 | 19 - 20 | 2408 | 70 | ||
XM_017016187.1 | XP_016871676.1 | 18 - 19 | 2207 | 70 | |
XM_017016188.1 | XP_016871677.1 | 18 - 19 | 2801 | 70 | |
XM_017016189.1 | XP_016871678.1 | 19 - 20 | 2200 | 70 | |
XM_017016190.1 | XP_016871679.1 | 18 - 19 | 2327 | 70 | |
GenBank mRNA | AB209204.1 | - | 9 - 10 | 933 | 70 |
AK093287.1 | - | 8 - 9 | 840 | 70 | |
AK302578.1 | - | 8 - 9 | 821 | 70 | |
AK312810.1 | - | 18 - 19 | 2209 | 70 | |
AK316407.1 | - | 8 - 9 | 840 | 70 | |
BC096336.1 | - | 18 - 19 | 2248 | 70 | |
BC096337.1 | - | 18 - 19 | 2248 | 70 | |
BC096339.3 | - | 18 - 19 | 2248 | 70 | |
BX648462.1 | - | 18 - 19 | 2449 | 70 | |
M21188.1 | - | 18 - 19 | 2265 | 70 |