Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031685.2 | NP_001026855.2 | 10 - 11 | 1632 | 81 |
NM_005426.2 | NP_005417.1 | 11 - 12 | 1764 | 81 | |
XM_011544268.1 | XP_011542570.1 | 9 - 10 | 1493 | 81 | |
XM_011544269.2 | XP_011542571.1 | 7 - 8 | 1151 | 81 | |
XM_017002220.1 | XP_016857709.1 | 11 - 12 | 1773 | 81 | |
GenBank mRNA | AJ318888.2 | - | 10 - 11 | 1577 | 81 |
AK294432.1 | - | 10 - 11 | 1484 | 81 | |
AK294964.1 | - | 3 - 4 | 652 | 81 | |
AK295378.1 | - | 4 - 5 | 356 | 81 | |
AK316016.1 | - | 3 - 4 | 554 | 81 | |
AK316103.1 | - | 3 - 4 | 532 | 81 | |
BC040247.1 | - | 8 - 9 | 1108 | 81 | |
BC046150.1 | - | 10 - 11 | 1575 | 81 | |
BC058918.1 | - | 10 - 11 | 1590 | 81 | |
U58334.1 | - | 11 - 12 | 1709 | 81 |