Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012311.3 | NP_036443.1 | 1 - 2 | 205 | 93 |
NR_045609.1 | 1 - 2 | 205 | 93 | ||
XM_006717434.3 | XP_006717497.1 | 1 - 2 | 200 | 93 | |
XM_011519427.1 | XP_011517729.1 | 1 - 2 | 200 | 93 | |
GenBank mRNA | AJ005273.1 | - | 1 - 2 | 176 | 93 |
AK301789.1 | - | 1 - 2 | 205 | 93 | |
AK309059.1 | - | 1 - 2 | 141 | 93 | |
BC017309.2 | - | 1 - 2 | 122 | 93 | |
BX504040.1 | - | 1 - 2 | 172 | 93 |