Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161619.1 | NP_001155091.1 | 1 - 2 | 192 | 73 |
NM_020175.2 | NP_064560.2 | 1 - 2 | 192 | 73 | |
XM_017027020.1 | XP_016882509.1 | 1 - 2 | 164 | 73 | |
GenBank mRNA | AK023958.1 | - | 1 - 2 | 117 | 73 |
AK225840.1 | - | 1 - 2 | 117 | 73 | |
AK297037.1 | - | 1 - 2 | 179 | 73 | |
AK315691.1 | - | 1 - 2 | 169 | 73 | |
AL365411.1 | - | 1 - 2 | 46 | 73 | |
BC004549.2 | - | 1 - 2 | 160 | 73 | |
BC009973.1 | - | 1 - 2 | 143 | 73 | |
DA839738.1 | - | 1 - 2 | 194 | 73 |