Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184891.1 | NP_001171820.1 | 2 - 3 | 330 | 73 |
NM_025207.4 | NP_079483.3 | 1 - 2 | 774 | 73 | |
NM_201398.2 | NP_958800.1 | 2 - 3 | 330 | 73 | |
GenBank mRNA | AF481877.1 | - | 1 - 2 | 82 | 73 |
AF520568.1 | - | 1 - 2 | 695 | 73 | |
AK025979.1 | - | 2 - 3 | 258 | 73 | |
AK090542.1 | - | 1 - 2 | 716 | 73 | |
AK308243.1 | - | 2 - 3 | 264 | 73 | |
BC011378.1 | - | 2 - 3 | 259 | 73 | |
BC021096.1 | - | 2 - 3 | 248 | 73 | |
BC032323.1 | - | 2 - 3 | 265 | 73 | |
BI919061.1 | - | 2 - 3 | 312 | 73 | |
DC371856.1 | - | 2 - 3 | 331 | 73 | |
DQ458779.1 | - | 1 - 2 | 373 | 73 |