Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145255.3 | NP_660298.2 | 2 - 3 | 252 | 68 |
NM_148887.2 | NP_683685.1 | 3 - 4 | 350 | 68 | |
NR_037575.1 | 2 - 3 | 401 | 68 | ||
GenBank mRNA | AK127167.1 | - | 2 - 3 | 1127 | 68 |
AK293624.1 | - | 2 - 3 | 390 | 68 | |
AL558270.3 | - | 3 - 4 | 315 | 68 | |
AL701268.1 | - | 2 - 3 | 249 | 68 | |
BC015904.1 | - | 2 - 3 | 223 | 68 | |
BC039852.1 | - | 2 - 3 | 231 | 68 | |
BC052601.1 | - | 2 - 3 | 241 | 68 | |
DA137043.1 | - | 2 - 3 | 252 | 68 |