Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13563073-13563316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000208 c.3958C>T p.R1320C Chr.12: 13563280-13563280 on Build GRCh38
6841620 c.4003C>T p.P1335S Chr.12: 13563235-13563235 on Build GRCh38
4517694 c.3993_3994GG>AA p.M1331_D1332>IN Chr.12: 13563244-13563245 on Build GRCh38
5591181 c.4108G>A p.G1370R Chr.12: 13563130-13563130 on Build GRCh38
4493330 c.4136C>T p.P1379L Chr.12: 13563102-13563102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371190262 Chr.12: 13563133-13563133 on Build GRCh38
rs202144479 Chr.12: 13563177-13563177 on Build GRCh38
rs200657400 Chr.12: 13563293-13563293 on Build GRCh38
rs1043559346 Chr.12: 13563136-13563136 on Build GRCh38
rs200255226 Chr.12: 13563274-13563274 on Build GRCh38
rs371762359 Chr.12: 13563217-13563217 on Build GRCh38
rs772733555 Chr.12: 13563198-13563198 on Build GRCh38
rs763372245 Chr.12: 13563189-13563189 on Build GRCh38
rs749867196 Chr.12: 13563110-13563110 on Build GRCh38
rs759438353 Chr.12: 13563207-13563207 on Build GRCh38
rs796052579 Chr.12: 13563172-13563172 on Build GRCh38
rs767675317 Chr.12: 13563129-13563129 on Build GRCh38
rs764080484 Chr.12: 13563230-13563230 on Build GRCh38
rs200021190 Chr.12: 13563134-13563134 on Build GRCh38
rs766929951 Chr.12: 13563098-13563098 on Build GRCh38
rs141730031 Chr.12: 13563281-13563281 on Build GRCh38
rs771307465 Chr.12: 13563146-13563146 on Build GRCh38
rs752473485 Chr.12: 13563221-13563221 on Build GRCh38
rs746083529 Chr.12: 13563246-13563246 on Build GRCh38
rs754738111 Chr.12: 13563130-13563130 on Build GRCh38
rs200421469 Chr.12: 13563244-13563244 on Build GRCh38
rs201670483 Chr.12: 13563257-13563257 on Build GRCh38
rs375217280 Chr.12: 13563161-13563161 on Build GRCh38
rs896420979 Chr.12: 13563250-13563250 on Build GRCh38
rs199803550 Chr.12: 13563292-13563292 on Build GRCh38
rs770040831 Chr.12: 13563162-13563162 on Build GRCh38
rs755797497 Chr.12: 13563126-13563126 on Build GRCh38
rs200035225 Chr.12: 13563245-13563245 on Build GRCh38
rs781529910 Chr.12: 13563160-13563160 on Build GRCh38
rs776247380 Chr.12: 13563203-13563203 on Build GRCh38
rs201214704 Chr.12: 13563200-13563200 on Build GRCh38
rs758288071 Chr.12: 13563224-13563224 on Build GRCh38
rs200217700 Chr.12: 13563124-13563124 on Build GRCh38
rs761812510 Chr.12: 13563180-13563182 on Build GRCh38
rs976298533 Chr.12: 13563123-13563123 on Build GRCh38
rs766060662 Chr.12: 13563202-13563202 on Build GRCh38
rs774808996 Chr.12: 13563287-13563287 on Build GRCh38
rs761443441 Chr.12: 13563096-13563096 on Build GRCh38
rs199803412 Chr.12: 13563194-13563194 on Build GRCh38
rs201732760 Chr.12: 13563131-13563131 on Build GRCh38
rs202102183 Chr.12: 13563216-13563216 on Build GRCh38
rs112932810 Chr.12: 13563137-13563137 on Build GRCh38
rs201335987 Chr.12: 13563229-13563229 on Build GRCh38
rs776130564 Chr.12: 13563168-13563168 on Build GRCh38
rs574536056 Chr.12: 13563226-13563226 on Build GRCh38
rs780293991 Chr.12: 13563258-13563258 on Build GRCh38
rs769147604 Chr.12: 13563197-13563197 on Build GRCh38
rs747033161 Chr.12: 13563140-13563140 on Build GRCh38
rs146792012 Chr.12: 13563125-13563125 on Build GRCh38
rs200604023 Chr.12: 13563223-13563223 on Build GRCh38

View Full Product Details