Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017934.5 | NP_060404.3 | 2 - 3 | 327 | 81 |
XM_005248729.4 | XP_005248786.1 | 2 - 3 | 327 | 81 | |
XM_011535919.1 | XP_011534221.1 | 2 - 3 | 327 | 81 | |
XM_017010989.1 | XP_016866478.1 | 2 - 3 | 315 | 81 | |
GenBank mRNA | AJ303102.1 | - | 2 - 3 | 114 | 81 |
AK057039.1 | - | 2 - 3 | 269 | 81 | |
BC021905.1 | - | 2 - 3 | 216 | 81 | |
BC137488.1 | - | 2 - 3 | 207 | 81 | |
DA500781.1 | - | 2 - 3 | 327 | 81 | |
DQ924532.1 | - | 2 - 3 | 101 | 81 |