Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86611403-86611664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955620 c.1633T>C p.Y545H Chr.8: 86611617-86611617 on Build GRCh38
7252479 c.1601A>G p.Y534C Chr.8: 86611649-86611649 on Build GRCh38
6365854 c.1627G>T p.V543F Chr.8: 86611623-86611623 on Build GRCh38
6364658 c.1621A>G p.K541E Chr.8: 86611629-86611629 on Build GRCh38
1458492 c.1600T>G p.Y534D Chr.8: 86611650-86611650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924653883 Chr.8: 86611517-86611517 on Build GRCh38
rs762516971 Chr.8: 86611553-86611553 on Build GRCh38
rs577987584 Chr.8: 86611488-86611488 on Build GRCh38
rs756606775 Chr.8: 86611564-86611564 on Build GRCh38
rs544416880 Chr.8: 86611459-86611459 on Build GRCh38
rs913186769 Chr.8: 86611614-86611614 on Build GRCh38
rs751218723 Chr.8: 86611500-86611500 on Build GRCh38
rs1042860696 Chr.8: 86611449-86611449 on Build GRCh38
rs746817996 Chr.8: 86611623-86611623 on Build GRCh38
rs373844401 Chr.8: 86611568-86611568 on Build GRCh38
rs897554395 Chr.8: 86611612-86611612 on Build GRCh38
rs767869910 Chr.8: 86611556-86611556 on Build GRCh38
rs914537784 Chr.8: 86611476-86611476 on Build GRCh38
rs527693877 Chr.8: 86611534-86611534 on Build GRCh38
rs777894511 Chr.8: 86611527-86611528 on Build GRCh38
rs375020827 Chr.8: 86611550-86611550 on Build GRCh38
rs758570863 Chr.8: 86611597-86611597 on Build GRCh38
rs370858784 Chr.8: 86611547-86611547 on Build GRCh38
rs747653530 Chr.8: 86611628-86611628 on Build GRCh38
rs28517318 Chr.8: 86611521-86611521 on Build GRCh38
rs865921625 Chr.8: 86611518-86611518 on Build GRCh38
rs776086758 Chr.8: 86611540-86611540 on Build GRCh38
rs757475831 Chr.8: 86611582-86611582 on Build GRCh38
rs763692214 Chr.8: 86611453-86611453 on Build GRCh38
rs28629931 Chr.8: 86611485-86611485 on Build GRCh38
rs753951890 Chr.8: 86611579-86611579 on Build GRCh38
rs368065564 Chr.8: 86611563-86611563 on Build GRCh38
rs772916629 Chr.8: 86611552-86611552 on Build GRCh38
rs140853266 Chr.8: 86611606-86611606 on Build GRCh38
rs183005285 Chr.8: 86611533-86611533 on Build GRCh38
rs35903042 Chr.8: 86611624-86611624 on Build GRCh38
rs977899634 Chr.8: 86611494-86611494 on Build GRCh38
rs1009610900 Chr.8: 86611437-86611437 on Build GRCh38
rs779134560 Chr.8: 86611586-86611586 on Build GRCh38
rs375040623 Chr.8: 86611529-86611529 on Build GRCh38
rs933144396 Chr.8: 86611557-86611557 on Build GRCh38
rs750611810 Chr.8: 86611596-86611596 on Build GRCh38
rs781166193 Chr.8: 86611627-86611627 on Build GRCh38

View Full Product Details