Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113416658-113416915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
UniGene
Hs.73893
Gene Chromosome Location
Chr.11: 113409595-113475279 on Build GRCh38
Transcripts
NM_000795.3 NM_016574.3 XM_017017296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4867005 c.517G>A p.G173R Chr.11: 113416878-113416878 on Build GRCh38
923194 c.517G>A p.G173R Chr.11: 113416878-113416878 on Build GRCh38
923195 c.513C>G p.L171L Chr.11: 113416882-113416882 on Build GRCh38
4551089 c.518G>A p.G173E Chr.11: 113416877-113416877 on Build GRCh38
3868452 c.500C>T p.S167F Chr.11: 113416895-113416895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780508433 Chr.11: 113416771-113416771 on Build GRCh38
rs769717591 Chr.11: 113416872-113416872 on Build GRCh38
rs776148708 Chr.11: 113416869-113416869 on Build GRCh38
rs745365313 Chr.11: 113416776-113416776 on Build GRCh38
rs147347002 Chr.11: 113416879-113416879 on Build GRCh38
rs1029522633 Chr.11: 113416800-113416800 on Build GRCh38
rs919231969 Chr.11: 113416772-113416772 on Build GRCh38
rs563056664 Chr.11: 113416804-113416804 on Build GRCh38
rs756934582 Chr.11: 113416694-113416694 on Build GRCh38
rs35352421 Chr.11: 113416789-113416789 on Build GRCh38
rs571869697 Chr.11: 113416866-113416866 on Build GRCh38
rs532004608 Chr.11: 113416807-113416807 on Build GRCh38
rs761794375 Chr.11: 113416871-113416871 on Build GRCh38
rs560570403 Chr.11: 113416717-113416717 on Build GRCh38
rs868115222 Chr.11: 113416768-113416768 on Build GRCh38
rs773041317 Chr.11: 113416878-113416878 on Build GRCh38
rs552009249 Chr.11: 113416861-113416861 on Build GRCh38
rs963965816 Chr.11: 113416747-113416747 on Build GRCh38
rs908309994 Chr.11: 113416820-113416820 on Build GRCh38
rs982609838 Chr.11: 113416806-113416806 on Build GRCh38
rs34482574 Chr.11: 113416683-113416684 on Build GRCh38
rs953542043 Chr.11: 113416702-113416702 on Build GRCh38
rs945097632 Chr.11: 113416875-113416875 on Build GRCh38
rs368285251 Chr.11: 113416835-113416835 on Build GRCh38
rs532435544 Chr.11: 113416849-113416849 on Build GRCh38
rs779932957 Chr.11: 113416819-113416819 on Build GRCh38
rs529250993 Chr.11: 113416766-113416766 on Build GRCh38
rs533098066 Chr.11: 113416867-113416867 on Build GRCh38
rs749272967 Chr.11: 113416845-113416845 on Build GRCh38
rs1032940771 Chr.11: 113416734-113416734 on Build GRCh38
rs749643059 Chr.11: 113416817-113416817 on Build GRCh38
rs986303957 Chr.11: 113416716-113416716 on Build GRCh38
rs966494813 Chr.11: 113416755-113416755 on Build GRCh38
rs540320645 Chr.11: 113416713-113416713 on Build GRCh38
rs926566118 Chr.11: 113416799-113416799 on Build GRCh38
rs35761620 Chr.11: 113416783-113416784 on Build GRCh38
rs200614179 Chr.11: 113416841-113416841 on Build GRCh38

View Full Product Details