Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67230279-67230504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370501 c.3037C>T p.R1013C Chr.16: 67230328-67230328 on Build GRCh38
4684671 c.2942G>A p.R981Q Chr.16: 67230423-67230423 on Build GRCh38
3387536 c.3016G>A p.E1006K Chr.16: 67230349-67230349 on Build GRCh38
128003 c.2977C>A p.R993R Chr.16: 67230388-67230388 on Build GRCh38
1679174 c.3012C>A p.Y1004* Chr.16: 67230353-67230353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769610007 Chr.16: 67230345-67230345 on Build GRCh38
rs116268850 Chr.16: 67230394-67230394 on Build GRCh38
rs766205021 Chr.16: 67230429-67230429 on Build GRCh38
rs935221389 Chr.16: 67230476-67230476 on Build GRCh38
rs780524217 Chr.16: 67230309-67230309 on Build GRCh38
rs144753004 Chr.16: 67230434-67230434 on Build GRCh38
rs553201423 Chr.16: 67230479-67230479 on Build GRCh38
rs373944685 Chr.16: 67230405-67230405 on Build GRCh38
rs749039758 Chr.16: 67230414-67230414 on Build GRCh38
rs762980260 Chr.16: 67230432-67230432 on Build GRCh38
rs561555976 Chr.16: 67230304-67230304 on Build GRCh38
rs764765882 Chr.16: 67230481-67230481 on Build GRCh38
rs754215765 Chr.16: 67230374-67230374 on Build GRCh38
rs569958308 Chr.16: 67230423-67230423 on Build GRCh38
rs768438495 Chr.16: 67230416-67230416 on Build GRCh38
rs768363225 Chr.16: 67230459-67230459 on Build GRCh38
rs746053817 Chr.16: 67230457-67230457 on Build GRCh38
rs747184984 Chr.16: 67230341-67230341 on Build GRCh38
rs751703092 Chr.16: 67230451-67230451 on Build GRCh38
rs373561996 Chr.16: 67230470-67230470 on Build GRCh38
rs773281297 Chr.16: 67230466-67230466 on Build GRCh38
rs762793829 Chr.16: 67230347-67230347 on Build GRCh38
rs376723898 Chr.16: 67230317-67230317 on Build GRCh38
rs779295641 Chr.16: 67230393-67230393 on Build GRCh38
rs763650649 Chr.16: 67230444-67230444 on Build GRCh38
rs200317614 Chr.16: 67230421-67230421 on Build GRCh38
rs769648476 Chr.16: 67230352-67230352 on Build GRCh38
rs138705012 Chr.16: 67230450-67230450 on Build GRCh38
rs772076551 Chr.16: 67230324-67230324 on Build GRCh38
rs951567787 Chr.16: 67230378-67230378 on Build GRCh38
rs758611946 Chr.16: 67230395-67230395 on Build GRCh38
rs376362919 Chr.16: 67230307-67230307 on Build GRCh38
rs781469513 Chr.16: 67230413-67230413 on Build GRCh38
rs190207170 Chr.16: 67230465-67230465 on Build GRCh38
rs772896045 Chr.16: 67230426-67230426 on Build GRCh38
rs766412779 Chr.16: 67230340-67230341 on Build GRCh38
rs778804997 Chr.16: 67230460-67230460 on Build GRCh38
rs766104188 Chr.16: 67230366-67230366 on Build GRCh38
rs777791137 Chr.16: 67230399-67230399 on Build GRCh38
rs775305958 Chr.16: 67230346-67230346 on Build GRCh38
rs998356242 Chr.16: 67230355-67230355 on Build GRCh38
rs528954834 Chr.16: 67230310-67230310 on Build GRCh38
rs781283797 Chr.16: 67230454-67230454 on Build GRCh38
rs773436492 Chr.16: 67230327-67230327 on Build GRCh38
rs759835715 Chr.16: 67230433-67230433 on Build GRCh38
rs747061959 Chr.16: 67230403-67230403 on Build GRCh38
rs760251884 Chr.16: 67230428-67230428 on Build GRCh38
rs367934888 Chr.16: 67230445-67230445 on Build GRCh38
rs559455218 Chr.16: 67230318-67230318 on Build GRCh38
rs761241527 Chr.16: 67230356-67230356 on Build GRCh38
rs752112353 Chr.16: 67230368-67230370 on Build GRCh38
rs370189170 Chr.16: 67230452-67230452 on Build GRCh38
rs766454816 Chr.16: 67230480-67230480 on Build GRCh38
rs755368508 Chr.16: 67230377-67230377 on Build GRCh38
rs775224846 Chr.16: 67230360-67230360 on Build GRCh38
rs140269091 Chr.16: 67230471-67230471 on Build GRCh38
rs755869068 Chr.16: 67230364-67230364 on Build GRCh38
rs530863029 Chr.16: 67230447-67230447 on Build GRCh38
rs201674604 Chr.16: 67230464-67230464 on Build GRCh38
rs1039772478 Chr.16: 67230351-67230351 on Build GRCh38
rs755490026 Chr.16: 67230316-67230316 on Build GRCh38

View Full Product Details