Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27548516-27548743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220411 c.1246A>G p.I416V Chr.9: 27548570-27548570 on Build GRCh38
1331593 c.1170C>T p.G390G Chr.9: 27548646-27548646 on Build GRCh38
4889779 c.1232C>G p.T411R Chr.9: 27548584-27548584 on Build GRCh38
6271074 c.1218C>A p.H406Q Chr.9: 27548598-27548598 on Build GRCh38
1108018 c.1254C>T p.D418D Chr.9: 27548562-27548562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991962885 Chr.9: 27548656-27548656 on Build GRCh38
rs759218702 Chr.9: 27548557-27548557 on Build GRCh38
rs749761510 Chr.9: 27548708-27548708 on Build GRCh38
rs541965231 Chr.9: 27548606-27548606 on Build GRCh38
rs758512117 Chr.9: 27548609-27548609 on Build GRCh38
rs376835503 Chr.9: 27548715-27548715 on Build GRCh38
rs769477910 Chr.9: 27548558-27548558 on Build GRCh38
rs2643806 Chr.9: 27548639-27548639 on Build GRCh38
rs776218894 Chr.9: 27548556-27548556 on Build GRCh38
rs750942348 Chr.9: 27548577-27548577 on Build GRCh38
rs34602100 Chr.9: 27548583-27548583 on Build GRCh38
rs201585198 Chr.9: 27548712-27548712 on Build GRCh38
rs780934681 Chr.9: 27548628-27548628 on Build GRCh38
rs770228283 Chr.9: 27548610-27548610 on Build GRCh38
rs773541376 Chr.9: 27548695-27548695 on Build GRCh38
rs811641 Chr.9: 27548612-27548612 on Build GRCh38
rs747091754 Chr.9: 27548688-27548689 on Build GRCh38
rs778862260 Chr.9: 27548617-27548618 on Build GRCh38
rs185358772 Chr.9: 27548632-27548632 on Build GRCh38
rs369969754 Chr.9: 27548682-27548682 on Build GRCh38
rs1037213590 Chr.9: 27548674-27548674 on Build GRCh38
rs760887239 Chr.9: 27548578-27548578 on Build GRCh38
rs201734930 Chr.9: 27548562-27548562 on Build GRCh38
rs767903441 Chr.9: 27548571-27548571 on Build GRCh38
rs374772194 Chr.9: 27548672-27548672 on Build GRCh38
rs182259442 Chr.9: 27548549-27548549 on Build GRCh38
rs775188943 Chr.9: 27548643-27548643 on Build GRCh38
rs762581307 Chr.9: 27548568-27548568 on Build GRCh38
rs755216584 Chr.9: 27548587-27548587 on Build GRCh38
rs199877251 Chr.9: 27548687-27548687 on Build GRCh38
rs779339076 Chr.9: 27548591-27548591 on Build GRCh38
rs376801304 Chr.9: 27548679-27548679 on Build GRCh38
rs530261789 Chr.9: 27548586-27548586 on Build GRCh38
rs746990973 Chr.9: 27548625-27548625 on Build GRCh38
rs939774089 Chr.9: 27548659-27548659 on Build GRCh38
rs745710266 Chr.9: 27548636-27548636 on Build GRCh38

View Full Product Details