Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13564219-13564478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871152 c.2933G>A p.S978N Chr.12: 13564305-13564305 on Build GRCh38
6209120 c.3005G>A p.G1002E Chr.12: 13564233-13564233 on Build GRCh38
343446 c.2815C>A p.R939S Chr.12: 13564423-13564423 on Build GRCh38
937464 c.2934C>T p.S978S Chr.12: 13564304-13564304 on Build GRCh38
4541004 c.2929G>A p.D977N Chr.12: 13564309-13564309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959107851 Chr.12: 13564359-13564359 on Build GRCh38
rs547507161 Chr.12: 13564382-13564382 on Build GRCh38
rs760531029 Chr.12: 13564241-13564241 on Build GRCh38
rs771565713 Chr.12: 13564391-13564391 on Build GRCh38
rs144956856 Chr.12: 13564279-13564279 on Build GRCh38
rs369299730 Chr.12: 13564415-13564415 on Build GRCh38
rs199677214 Chr.12: 13564458-13564458 on Build GRCh38
rs772837930 Chr.12: 13564238-13564238 on Build GRCh38
rs147956755 Chr.12: 13564307-13564307 on Build GRCh38
rs77493389 Chr.12: 13564430-13564430 on Build GRCh38
rs778202551 Chr.12: 13564300-13564300 on Build GRCh38
rs755495386 Chr.12: 13564287-13564292 on Build GRCh38
rs781740648 Chr.12: 13564370-13564372 on Build GRCh38
rs150445188 Chr.12: 13564425-13564425 on Build GRCh38
rs747235156 Chr.12: 13564301-13564301 on Build GRCh38
rs201982602 Chr.12: 13564393-13564393 on Build GRCh38
rs766092483 Chr.12: 13564247-13564247 on Build GRCh38
rs199941557 Chr.12: 13564447-13564447 on Build GRCh38
rs201085200 Chr.12: 13564448-13564448 on Build GRCh38
rs376328340 Chr.12: 13564377-13564377 on Build GRCh38
rs765183831 Chr.12: 13564278-13564278 on Build GRCh38
rs876661102 Chr.12: 13564310-13564312 on Build GRCh38
rs200724001 Chr.12: 13564413-13564413 on Build GRCh38
rs764278787 Chr.12: 13564459-13564459 on Build GRCh38
rs200538130 Chr.12: 13564357-13564357 on Build GRCh38
rs202006928 Chr.12: 13564265-13564265 on Build GRCh38
rs758533546 Chr.12: 13564292-13564292 on Build GRCh38
rs969024709 Chr.12: 13564240-13564240 on Build GRCh38
rs753613072 Chr.12: 13564249-13564249 on Build GRCh38
rs886049101 Chr.12: 13564372-13564372 on Build GRCh38
rs747714077 Chr.12: 13564383-13564383 on Build GRCh38
rs776323617 Chr.12: 13564422-13564422 on Build GRCh38
rs201252525 Chr.12: 13564328-13564328 on Build GRCh38
rs926331767 Chr.12: 13564332-13564332 on Build GRCh38
rs754896277 Chr.12: 13564258-13564258 on Build GRCh38
rs796052574 Chr.12: 13564302-13564302 on Build GRCh38
rs200696950 Chr.12: 13564325-13564325 on Build GRCh38
rs886049100 Chr.12: 13564348-13564348 on Build GRCh38
rs199536073 Chr.12: 13564331-13564331 on Build GRCh38
rs140573925 Chr.12: 13564412-13564412 on Build GRCh38

View Full Product Details