Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27920500-27920725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Gene Symbol
PDX1-AS1
Entrez Gene ID
100861550
Gene Name
PDX1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.416201
Gene Chromosome Location
Chr.13: 27829759-27921404 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2070076 c.385G>A p.A129T Chr.13: 27920523-27920523 on Build GRCh38
4596091 c.401G>A p.W134* Chr.13: 27920539-27920539 on Build GRCh38
5702794 c.395G>A p.G132D Chr.13: 27920533-27920533 on Build GRCh38
4596092 c.402G>A p.W134* Chr.13: 27920540-27920540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991097151 Chr.13: 27920658-27920658 on Build GRCh38
rs745673876 Chr.13: 27920592-27920592 on Build GRCh38
rs949580886 Chr.13: 27920678-27920678 on Build GRCh38
rs747580950 Chr.13: 27920594-27920594 on Build GRCh38
rs557040229 Chr.13: 27920617-27920617 on Build GRCh38
rs756308187 Chr.13: 27920539-27920539 on Build GRCh38
rs1037295010 Chr.13: 27920659-27920659 on Build GRCh38
rs780282811 Chr.13: 27920544-27920544 on Build GRCh38
rs770779149 Chr.13: 27920585-27920585 on Build GRCh38
rs939854346 Chr.13: 27920639-27920639 on Build GRCh38
rs573487482 Chr.13: 27920642-27920642 on Build GRCh38
rs539662722 Chr.13: 27920635-27920635 on Build GRCh38
rs757558416 Chr.13: 27920533-27920533 on Build GRCh38
rs901256901 Chr.13: 27920668-27920668 on Build GRCh38
rs750762696 Chr.13: 27920537-27920537 on Build GRCh38
rs1020690285 Chr.13: 27920558-27920558 on Build GRCh38
rs982604102 Chr.13: 27920679-27920679 on Build GRCh38
rs776429718 Chr.13: 27920588-27920588 on Build GRCh38
rs749338559 Chr.13: 27920555-27920555 on Build GRCh38
rs755101362 Chr.13: 27920569-27920569 on Build GRCh38
rs879018495 Chr.13: 27920618-27920618 on Build GRCh38
rs757073644 Chr.13: 27920693-27920693 on Build GRCh38
rs956251210 Chr.13: 27920551-27920551 on Build GRCh38
rs149360955 Chr.13: 27920670-27920670 on Build GRCh38
rs200827032 Chr.13: 27920577-27920577 on Build GRCh38
rs938296762 Chr.13: 27920667-27920667 on Build GRCh38
rs544041273 Chr.13: 27920619-27920619 on Build GRCh38
rs752168064 Chr.13: 27920532-27920532 on Build GRCh38
rs781552927 Chr.13: 27920534-27920534 on Build GRCh38
rs970907657 Chr.13: 27920576-27920576 on Build GRCh38
rs967027661 Chr.13: 27920604-27920604 on Build GRCh38
rs1009031133 Chr.13: 27920561-27920561 on Build GRCh38
rs373337234 Chr.13: 27920614-27920614 on Build GRCh38
rs926283932 Chr.13: 27920599-27920599 on Build GRCh38
rs769449809 Chr.13: 27920594-27920594 on Build GRCh38
rs553135605 Chr.13: 27920637-27920637 on Build GRCh38
rs747038170 Chr.13: 27920578-27920578 on Build GRCh38
rs908002051 Chr.13: 27920688-27920688 on Build GRCh38

View Full Product Details