Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42745871-42746142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4808172 c.3566A>G p.K1189R Chr.15: 42745964-42745964 on Build GRCh38
6810857 c.3444C>T p.R1148R Chr.15: 42746086-42746086 on Build GRCh38
6201356 c.3510G>A p.R1170R Chr.15: 42746020-42746020 on Build GRCh38
84286 c.3399T>C p.S1133S Chr.15: 42746131-42746131 on Build GRCh38
4901648 c.3392C>T p.P1131L Chr.15: 42746138-42746138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769062274 Chr.15: 42746100-42746100 on Build GRCh38
rs748778273 Chr.15: 42746063-42746063 on Build GRCh38
rs770444240 Chr.15: 42746064-42746064 on Build GRCh38
rs538828713 Chr.15: 42746004-42746004 on Build GRCh38
rs372524749 Chr.15: 42745965-42745965 on Build GRCh38
rs751381694 Chr.15: 42745960-42745960 on Build GRCh38
rs373828657 Chr.15: 42746013-42746013 on Build GRCh38
rs201674605 Chr.15: 42746042-42746042 on Build GRCh38
rs1064028 Chr.15: 42746008-42746008 on Build GRCh38
rs200891264 Chr.15: 42746045-42746045 on Build GRCh38
rs376907182 Chr.15: 42746041-42746041 on Build GRCh38
rs771394738 Chr.15: 42745890-42745890 on Build GRCh38
rs920391448 Chr.15: 42746002-42746002 on Build GRCh38
rs764592156 Chr.15: 42746015-42746015 on Build GRCh38
rs200469913 Chr.15: 42745991-42745991 on Build GRCh38
rs775998719 Chr.15: 42746091-42746091 on Build GRCh38
rs371070916 Chr.15: 42746059-42746059 on Build GRCh38
rs201347313 Chr.15: 42746110-42746110 on Build GRCh38
rs777708456 Chr.15: 42746053-42746053 on Build GRCh38
rs768169944 Chr.15: 42745903-42745903 on Build GRCh38
rs779134989 Chr.15: 42745957-42745957 on Build GRCh38
rs753222805 Chr.15: 42745909-42745909 on Build GRCh38
rs759930908 Chr.15: 42745899-42745899 on Build GRCh38
rs761907261 Chr.15: 42746032-42746032 on Build GRCh38
rs757372218 Chr.15: 42746003-42746003 on Build GRCh38
rs776195390 Chr.15: 42746012-42746012 on Build GRCh38
rs748766632 Chr.15: 42745988-42745988 on Build GRCh38
rs948463594 Chr.15: 42746049-42746049 on Build GRCh38
rs754153837 Chr.15: 42745945-42745945 on Build GRCh38
rs747336557 Chr.15: 42746097-42746097 on Build GRCh38
rs986629295 Chr.15: 42745914-42745914 on Build GRCh38
rs369214005 Chr.15: 42745964-42745964 on Build GRCh38
rs770503390 Chr.15: 42745989-42745989 on Build GRCh38
rs765413012 Chr.15: 42746036-42746036 on Build GRCh38
rs772540637 Chr.15: 42746005-42746005 on Build GRCh38
rs761842533 Chr.15: 42746105-42746105 on Build GRCh38
rs574340489 Chr.15: 42746088-42746088 on Build GRCh38
rs769405212 Chr.15: 42745977-42745977 on Build GRCh38
rs1017700372 Chr.15: 42745929-42745929 on Build GRCh38
rs35504571 Chr.15: 42746006-42746006 on Build GRCh38
rs199594294 Chr.15: 42746087-42746087 on Build GRCh38
rs777159553 Chr.15: 42746103-42746103 on Build GRCh38
rs199635198 Chr.15: 42745987-42745987 on Build GRCh38
rs576737530 Chr.15: 42746112-42746112 on Build GRCh38
rs757421014 Chr.15: 42745955-42745955 on Build GRCh38
rs375039510 Chr.15: 42745934-42745934 on Build GRCh38
rs35576058 Chr.15: 42745958-42745958 on Build GRCh38
rs200437800 Chr.15: 42746046-42746046 on Build GRCh38
rs753873211 Chr.15: 42746031-42746031 on Build GRCh38
rs983647656 Chr.15: 42746023-42746023 on Build GRCh38
rs774584978 Chr.15: 42745894-42745894 on Build GRCh38

View Full Product Details