Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74922729-74922990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOP2
Entrez Gene ID
92736
Gene Name
otopetrin 2
Gene Aliases
-
UniGene
Hs.352515
Gene Chromosome Location
Chr.17: 74922963-74933913 on Build GRCh38
Transcripts
XM_011525479.1 NM_001282489.2 NM_173477.4 XM_011524296.2

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
XM_011525479.1 NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5181377 c.105_106insC p.T36fs*22 Chr.17: 74922968-74922969 on Build GRCh38
6259780 c.162C>A p.R54R Chr.17: 74922912-74922912 on Build GRCh38
7396938 c.133C>A p.L45I Chr.17: 74922941-74922941 on Build GRCh38
6556659 c.124C>A p.H42N Chr.17: 74922950-74922950 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776132786 Chr.17: 74922878-74922878 on Build GRCh38
rs769657993 Chr.17: 74922929-74922929 on Build GRCh38
rs886042306 Chr.17: 74922923-74922923 on Build GRCh38
rs917200314 Chr.17: 74922928-74922928 on Build GRCh38
rs185144732 Chr.17: 74922792-74922792 on Build GRCh38
rs777746284 Chr.17: 74922921-74922921 on Build GRCh38
rs929326559 Chr.17: 74922939-74922939 on Build GRCh38
rs976207617 Chr.17: 74922902-74922902 on Build GRCh38
rs104894652 Chr.17: 74922961-74922961 on Build GRCh38
rs943856706 Chr.17: 74922888-74922888 on Build GRCh38
rs999584634 Chr.17: 74922966-74922966 on Build GRCh38
rs201967671 Chr.17: 74922938-74922938 on Build GRCh38
rs912421628 Chr.17: 74922854-74922854 on Build GRCh38
rs761731473 Chr.17: 74922879-74922879 on Build GRCh38
rs1007090577 Chr.17: 74922776-74922776 on Build GRCh38
rs979041144 Chr.17: 74922810-74922810 on Build GRCh38
rs397517926 Chr.17: 74922911-74922911 on Build GRCh38
rs104894651 Chr.17: 74922931-74922931 on Build GRCh38
rs768399553 Chr.17: 74922875-74922875 on Build GRCh38
rs746466465 Chr.17: 74922862-74922862 on Build GRCh38
rs1049056445 Chr.17: 74922969-74922969 on Build GRCh38
rs111801414 Chr.17: 74922817-74922817 on Build GRCh38
rs113248968 Chr.17: 74922899-74922899 on Build GRCh38
rs762948482 Chr.17: 74922944-74922944 on Build GRCh38
rs1054315517 Chr.17: 74922970-74922970 on Build GRCh38
rs45562933 Chr.17: 74922831-74922831 on Build GRCh38
rs985343878 Chr.17: 74922845-74922845 on Build GRCh38
rs976357763 Chr.17: 74922811-74922811 on Build GRCh38

View Full Product Details