Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18122143-18122400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC1
Entrez Gene ID
378884
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
Gene Aliases
EPM2A, EPM2B, MALIN, bA204B7.2
UniGene
Hs.348351
Gene Chromosome Location
Chr.6: 18120487-18122620 on Build GRCh38
Transcripts
NM_198586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455469 c.435C>T p.H145H Chr.6: 18122172-18122172 on Build GRCh38
5466769 c.417G>A p.G139G Chr.6: 18122190-18122190 on Build GRCh38
1076366 c.363C>A p.G121G Chr.6: 18122244-18122244 on Build GRCh38
6535869 c.463G>C p.D155H Chr.6: 18122144-18122144 on Build GRCh38
7282497 c.424G>A p.V142M Chr.6: 18122183-18122183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912302534 Chr.6: 18122357-18122357 on Build GRCh38
rs950907157 Chr.6: 18122230-18122230 on Build GRCh38
rs967125703 Chr.6: 18122216-18122216 on Build GRCh38
rs143537405 Chr.6: 18122185-18122185 on Build GRCh38
rs1011706675 Chr.6: 18122177-18122177 on Build GRCh38
rs568131096 Chr.6: 18122300-18122300 on Build GRCh38
rs756365176 Chr.6: 18122235-18122235 on Build GRCh38
rs750465793 Chr.6: 18122221-18122221 on Build GRCh38
rs370292624 Chr.6: 18122193-18122193 on Build GRCh38
rs986573010 Chr.6: 18122339-18122339 on Build GRCh38
rs148035405 Chr.6: 18122189-18122189 on Build GRCh38
rs886061254 Chr.6: 18122210-18122210 on Build GRCh38
rs10949483 Chr.6: 18122275-18122275 on Build GRCh38
rs755232826 Chr.6: 18122373-18122373 on Build GRCh38
rs759287189 Chr.6: 18122184-18122184 on Build GRCh38
rs764038720 Chr.6: 18122198-18122198 on Build GRCh38
rs770608651 Chr.6: 18122181-18122181 on Build GRCh38
rs753103968 Chr.6: 18122380-18122380 on Build GRCh38
rs200559475 Chr.6: 18122169-18122169 on Build GRCh38
rs762766683 Chr.6: 18122301-18122301 on Build GRCh38
rs754279385 Chr.6: 18122247-18122247 on Build GRCh38
rs774953311 Chr.6: 18122172-18122172 on Build GRCh38
rs769497255 Chr.6: 18122297-18122297 on Build GRCh38
rs780082503 Chr.6: 18122246-18122246 on Build GRCh38
rs754156735 Chr.6: 18122369-18122369 on Build GRCh38
rs187783545 Chr.6: 18122304-18122304 on Build GRCh38
rs779482781 Chr.6: 18122378-18122378 on Build GRCh38
rs376655413 Chr.6: 18122183-18122183 on Build GRCh38
rs774110305 Chr.6: 18122307-18122307 on Build GRCh38
rs761706418 Chr.6: 18122209-18122209 on Build GRCh38
rs769301934 Chr.6: 18122171-18122171 on Build GRCh38
rs570505924 Chr.6: 18122270-18122270 on Build GRCh38
rs761806425 Chr.6: 18122336-18122336 on Build GRCh38
rs963940421 Chr.6: 18122291-18122291 on Build GRCh38
rs893057832 Chr.6: 18122383-18122383 on Build GRCh38
rs113474848 Chr.6: 18122266-18122266 on Build GRCh38
rs767291745 Chr.6: 18122342-18122342 on Build GRCh38
rs909097062 Chr.6: 18122294-18122294 on Build GRCh38
rs767683762 Chr.6: 18122217-18122217 on Build GRCh38
rs750530014 Chr.6: 18122351-18122351 on Build GRCh38
rs779392254 Chr.6: 18122271-18122271 on Build GRCh38
rs796052758 Chr.6: 18122381-18122381 on Build GRCh38
rs766657432 Chr.6: 18122368-18122368 on Build GRCh38
rs774955028 Chr.6: 18122299-18122299 on Build GRCh38
rs796946441 Chr.6: 18122252-18122252 on Build GRCh38
rs772440533 Chr.6: 18122286-18122286 on Build GRCh38
rs115931931 Chr.6: 18122295-18122295 on Build GRCh38
rs760624567 Chr.6: 18122367-18122367 on Build GRCh38
rs751457173 Chr.6: 18122206-18122206 on Build GRCh38
rs371218834 Chr.6: 18122353-18122353 on Build GRCh38
rs796052755 Chr.6: 18122288-18122288 on Build GRCh38
rs758945379 Chr.6: 18122382-18122382 on Build GRCh38

View Full Product Details