Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52317600-52317867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940674 c.695C>T p.A232V Chr.12: 52317736-52317736 on Build GRCh38
6698283 c.683C>T p.S228L Chr.12: 52317748-52317748 on Build GRCh38
940673 c.699C>T p.N233N Chr.12: 52317732-52317732 on Build GRCh38
7269662 c.700G>A p.V234M Chr.12: 52317731-52317731 on Build GRCh38
5375392 c.694G>A p.A232T Chr.12: 52317737-52317737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758060854 Chr.12: 52317648-52317649 on Build GRCh38
rs779923045 Chr.12: 52317802-52317805 on Build GRCh38
rs574610233 Chr.12: 52317662-52317662 on Build GRCh38
rs149581248 Chr.12: 52317764-52317764 on Build GRCh38
rs746416217 Chr.12: 52317731-52317731 on Build GRCh38
rs780683896 Chr.12: 52317740-52317740 on Build GRCh38
rs373044241 Chr.12: 52317754-52317754 on Build GRCh38
rs754271207 Chr.12: 52317642-52317643 on Build GRCh38
rs749710516 Chr.12: 52317749-52317749 on Build GRCh38
rs916365094 Chr.12: 52317846-52317846 on Build GRCh38
rs751718404 Chr.12: 52317708-52317708 on Build GRCh38
rs1022097989 Chr.12: 52317680-52317680 on Build GRCh38
rs757384136 Chr.12: 52317796-52317796 on Build GRCh38
rs2248619 Chr.12: 52317666-52317666 on Build GRCh38
rs764886344 Chr.12: 52317785-52317785 on Build GRCh38
rs771505177 Chr.12: 52317820-52317820 on Build GRCh38
rs779875995 Chr.12: 52317734-52317734 on Build GRCh38
rs1011164040 Chr.12: 52317676-52317676 on Build GRCh38
rs377136758 Chr.12: 52317694-52317694 on Build GRCh38
rs866784254 Chr.12: 52317658-52317658 on Build GRCh38
rs770911936 Chr.12: 52317664-52317664 on Build GRCh38
rs781068595 Chr.12: 52317715-52317715 on Build GRCh38
rs780773670 Chr.12: 52317821-52317821 on Build GRCh38
rs953831263 Chr.12: 52317760-52317760 on Build GRCh38
rs781352531 Chr.12: 52317805-52317805 on Build GRCh38
rs2852460 Chr.12: 52317632-52317632 on Build GRCh38
rs761449359 Chr.12: 52317687-52317687 on Build GRCh38
rs961008827 Chr.12: 52317766-52317766 on Build GRCh38
rs375126673 Chr.12: 52317784-52317784 on Build GRCh38
rs753770238 Chr.12: 52317777-52317777 on Build GRCh38
rs746304771 Chr.12: 52317670-52317670 on Build GRCh38
rs775955586 Chr.12: 52317674-52317674 on Build GRCh38
rs766345093 Chr.12: 52317771-52317771 on Build GRCh38
rs749798679 Chr.12: 52317823-52317823 on Build GRCh38
rs755992731 Chr.12: 52317635-52317635 on Build GRCh38
rs778124250 Chr.12: 52317706-52317706 on Build GRCh38
rs770285501 Chr.12: 52317673-52317673 on Build GRCh38
rs766187594 Chr.12: 52317695-52317695 on Build GRCh38
rs780752334 Chr.12: 52317645-52317645 on Build GRCh38
rs972652254 Chr.12: 52317781-52317781 on Build GRCh38
rs117028083 Chr.12: 52317806-52317806 on Build GRCh38
rs916048349 Chr.12: 52317816-52317816 on Build GRCh38
rs2857667 Chr.12: 52317765-52317765 on Build GRCh38
rs553491257 Chr.12: 52317686-52317686 on Build GRCh38
rs748403427 Chr.12: 52317646-52317646 on Build GRCh38
rs750385511 Chr.12: 52317689-52317689 on Build GRCh38
rs751813981 Chr.12: 52317790-52317790 on Build GRCh38
rs761312848 Chr.12: 52317759-52317759 on Build GRCh38
rs35353807 Chr.12: 52317769-52317770 on Build GRCh38
rs112016211 Chr.12: 52317630-52317630 on Build GRCh38
rs2857666 Chr.12: 52317840-52317840 on Build GRCh38
rs757372724 Chr.12: 52317709-52317709 on Build GRCh38
rs753686095 Chr.12: 52317696-52317696 on Build GRCh38
rs550808670 Chr.12: 52317631-52317631 on Build GRCh38
rs758573053 Chr.12: 52317660-52317660 on Build GRCh38
rs762716889 Chr.12: 52317681-52317681 on Build GRCh38
rs186866078 Chr.12: 52317732-52317732 on Build GRCh38
rs192947508 Chr.12: 52317755-52317755 on Build GRCh38
rs202232721 Chr.12: 52317639-52317639 on Build GRCh38

View Full Product Details