Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23415644-23415893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MHRT
Entrez Gene ID
104564225
Gene Name
myosin heavy chain-associated RNA transcript
Gene Aliases
Myheart
UniGene
Gene Chromosome Location
Chr.14: 23415450-23417595 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
954741 c.5111A>G p.Q1704R Chr.14: 23415675-23415675 on Build GRCh38
1258461 c.5026C>T p.R1676W Chr.14: 23415760-23415760 on Build GRCh38
245863 c.5030G>A p.R1677H Chr.14: 23415756-23415756 on Build GRCh38
954742 c.5093C>T p.S1698F Chr.14: 23415693-23415693 on Build GRCh38
5802697 c.4984C>T p.R1662C Chr.14: 23415802-23415802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs727504844 Chr.14: 23415676-23415676 on Build GRCh38
rs797044602 Chr.14: 23415669-23415669 on Build GRCh38
rs111468196 Chr.14: 23415835-23415835 on Build GRCh38
rs570732103 Chr.14: 23415707-23415707 on Build GRCh38
rs753021990 Chr.14: 23415809-23415809 on Build GRCh38
rs730880915 Chr.14: 23415721-23415721 on Build GRCh38
rs768580382 Chr.14: 23415724-23415724 on Build GRCh38
rs768774626 Chr.14: 23415776-23415776 on Build GRCh38
rs377461670 Chr.14: 23415757-23415757 on Build GRCh38
rs752021154 Chr.14: 23415754-23415754 on Build GRCh38
rs397516239 Chr.14: 23415671-23415671 on Build GRCh38
rs45620235 Chr.14: 23415781-23415781 on Build GRCh38
rs763538103 Chr.14: 23415794-23415794 on Build GRCh38
rs762398027 Chr.14: 23415750-23415750 on Build GRCh38
rs753907340 Chr.14: 23415769-23415769 on Build GRCh38
rs397516234 Chr.14: 23415766-23415771 on Build GRCh38
rs747993770 Chr.14: 23415790-23415790 on Build GRCh38
rs878853838 Chr.14: 23415673-23415673 on Build GRCh38
rs200457830 Chr.14: 23415860-23415860 on Build GRCh38
rs750013359 Chr.14: 23415681-23415681 on Build GRCh38
rs3729830 Chr.14: 23415680-23415680 on Build GRCh38
rs730880811 Chr.14: 23415666-23415666 on Build GRCh38
rs769621446 Chr.14: 23415746-23415746 on Build GRCh38
rs869225593 Chr.14: 23415662-23415662 on Build GRCh38
rs747900941 Chr.14: 23415863-23415863 on Build GRCh38
rs773977507 Chr.14: 23415802-23415802 on Build GRCh38
rs772745454 Chr.14: 23415747-23415747 on Build GRCh38
rs374289523 Chr.14: 23415812-23415812 on Build GRCh38
rs766831916 Chr.14: 23415696-23415696 on Build GRCh38
rs761344083 Chr.14: 23415677-23415677 on Build GRCh38
rs763791649 Chr.14: 23415667-23415667 on Build GRCh38
rs761548214 Chr.14: 23415733-23415733 on Build GRCh38
rs776639582 Chr.14: 23415732-23415732 on Build GRCh38
rs554882421 Chr.14: 23415856-23415856 on Build GRCh38
rs797044601 Chr.14: 23415799-23415799 on Build GRCh38
rs397516235 Chr.14: 23415766-23415766 on Build GRCh38
rs753115999 Chr.14: 23415760-23415760 on Build GRCh38
rs748373063 Chr.14: 23415712-23415712 on Build GRCh38
rs267603953 Chr.14: 23415811-23415811 on Build GRCh38
rs550015662 Chr.14: 23415772-23415772 on Build GRCh38
rs730880914 Chr.14: 23415756-23415756 on Build GRCh38
rs779978846 Chr.14: 23415773-23415773 on Build GRCh38
rs781334634 Chr.14: 23415710-23415710 on Build GRCh38
rs146778113 Chr.14: 23415690-23415690 on Build GRCh38
rs587779394 Chr.14: 23415779-23415781 on Build GRCh38
rs769528619 Chr.14: 23415793-23415793 on Build GRCh38
rs373219734 Chr.14: 23415698-23415698 on Build GRCh38
rs45464193 Chr.14: 23415715-23415715 on Build GRCh38
rs962469283 Chr.14: 23415837-23415837 on Build GRCh38
rs535525636 Chr.14: 23415851-23415851 on Build GRCh38
rs370328209 Chr.14: 23415801-23415801 on Build GRCh38
rs1006534868 Chr.14: 23415807-23415807 on Build GRCh38
rs781115033 Chr.14: 23415782-23415782 on Build GRCh38
rs869034639 Chr.14: 23415674-23415674 on Build GRCh38
rs1057518857 Chr.14: 23415664-23415664 on Build GRCh38
rs764844610 Chr.14: 23415716-23415716 on Build GRCh38
rs772008016 Chr.14: 23415720-23415720 on Build GRCh38
rs727505070 Chr.14: 23415846-23415846 on Build GRCh38
rs369261897 Chr.14: 23415866-23415866 on Build GRCh38
rs727505033 Chr.14: 23415707-23415719 on Build GRCh38
rs2754153 Chr.14: 23415770-23415770 on Build GRCh38
rs397516233 Chr.14: 23415832-23415832 on Build GRCh38
rs149904793 Chr.14: 23415873-23415873 on Build GRCh38
rs766855849 Chr.14: 23415752-23415752 on Build GRCh38
rs139518691 Chr.14: 23415767-23415767 on Build GRCh38
rs886039010 Chr.14: 23415821-23415821 on Build GRCh38
rs786204381 Chr.14: 23415774-23415774 on Build GRCh38
rs112943492 Chr.14: 23415806-23415806 on Build GRCh38

View Full Product Details