Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173910733-173910958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899815 c.694G>A p.D232N Chr.1: 173910822-173910822 on Build GRCh38
3705252 c.664G>A p.V222M Chr.1: 173910852-173910852 on Build GRCh38
7250756 c.721G>A p.E241K Chr.1: 173910795-173910795 on Build GRCh38
6476288 c.654C>A p.I218I Chr.1: 173910862-173910862 on Build GRCh38
6577495 c.662G>C p.W221S Chr.1: 173910854-173910854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121909572 Chr.1: 173910849-173910849 on Build GRCh38
rs751357562 Chr.1: 173910768-173910768 on Build GRCh38
rs483352854 Chr.1: 173910875-173910875 on Build GRCh38
rs747558106 Chr.1: 173910924-173910924 on Build GRCh38
rs769820027 Chr.1: 173910785-173910785 on Build GRCh38
rs773688316 Chr.1: 173910919-173910919 on Build GRCh38
rs769911947 Chr.1: 173910810-173910810 on Build GRCh38
rs757200092 Chr.1: 173910775-173910775 on Build GRCh38
rs757009890 Chr.1: 173910856-173910856 on Build GRCh38
rs768914754 Chr.1: 173910803-173910803 on Build GRCh38
rs773814831 Chr.1: 173910822-173910822 on Build GRCh38
rs749510661 Chr.1: 173910800-173910800 on Build GRCh38
rs146733468 Chr.1: 173910846-173910846 on Build GRCh38
rs993457876 Chr.1: 173910755-173910755 on Build GRCh38
rs371099470 Chr.1: 173910934-173910934 on Build GRCh38
rs543358196 Chr.1: 173910897-173910897 on Build GRCh38
rs140452859 Chr.1: 173910823-173910823 on Build GRCh38
rs772710258 Chr.1: 173910812-173910812 on Build GRCh38
rs750413071 Chr.1: 173910869-173910869 on Build GRCh38
rs779121500 Chr.1: 173910904-173910904 on Build GRCh38
rs200861147 Chr.1: 173910797-173910797 on Build GRCh38
rs572252151 Chr.1: 173910784-173910784 on Build GRCh38
rs863224366 Chr.1: 173910871-173910871 on Build GRCh38
rs745970800 Chr.1: 173910760-173910760 on Build GRCh38
rs773408103 Chr.1: 173910899-173910899 on Build GRCh38
rs774877455 Chr.1: 173910938-173910938 on Build GRCh38
rs144084678 Chr.1: 173910767-173910767 on Build GRCh38
rs1018454989 Chr.1: 173910766-173910766 on Build GRCh38
rs267598176 Chr.1: 173910891-173910891 on Build GRCh38
rs879067636 Chr.1: 173910927-173910927 on Build GRCh38
rs767468032 Chr.1: 173910868-173910868 on Build GRCh38
rs121909571 Chr.1: 173910861-173910861 on Build GRCh38
rs756016928 Chr.1: 173910882-173910882 on Build GRCh38
rs772515943 Chr.1: 173910914-173910914 on Build GRCh38
rs774632265 Chr.1: 173910808-173910808 on Build GRCh38
rs113396092 Chr.1: 173910894-173910894 on Build GRCh38
rs775062927 Chr.1: 173910830-173910830 on Build GRCh38
rs2227627 Chr.1: 173910838-173910838 on Build GRCh38
rs558726250 Chr.1: 173910778-173910778 on Build GRCh38
rs968019175 Chr.1: 173910867-173910867 on Build GRCh38
rs368419985 Chr.1: 173910809-173910809 on Build GRCh38
rs749583360 Chr.1: 173910847-173910847 on Build GRCh38
rs748553880 Chr.1: 173910907-173910907 on Build GRCh38
rs780062808 Chr.1: 173910883-173910883 on Build GRCh38

View Full Product Details