Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50945344-50945605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086446 c.585C>G p.F195L Chr.13: 50945501-50945501 on Build GRCh38
6767098 c.562C>T p.R188W Chr.13: 50945478-50945478 on Build GRCh38
947930 c.549G>A p.V183V Chr.13: 50945465-50945465 on Build GRCh38
5983005 c.511G>T p.V171F Chr.13: 50945427-50945427 on Build GRCh38
6767101 c.563G>A p.R188Q Chr.13: 50945479-50945479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765643940 Chr.13: 50945483-50945489 on Build GRCh38
rs755648468 Chr.13: 50945528-50945528 on Build GRCh38
rs74555752 Chr.13: 50945470-50945470 on Build GRCh38
rs985375667 Chr.13: 50945396-50945396 on Build GRCh38
rs760862553 Chr.13: 50945465-50945465 on Build GRCh38
rs746658516 Chr.13: 50945384-50945384 on Build GRCh38
rs780872918 Chr.13: 50945492-50945492 on Build GRCh38
rs141049110 Chr.13: 50945439-50945439 on Build GRCh38
rs976988016 Chr.13: 50945554-50945554 on Build GRCh38
rs75184679 Chr.13: 50945445-50945445 on Build GRCh38
rs1027265653 Chr.13: 50945369-50945369 on Build GRCh38
rs776365023 Chr.13: 50945404-50945407 on Build GRCh38
rs902917241 Chr.13: 50945437-50945437 on Build GRCh38
rs896102356 Chr.13: 50945527-50945527 on Build GRCh38
rs763470120 Chr.13: 50945378-50945379 on Build GRCh38
rs1005947039 Chr.13: 50945524-50945524 on Build GRCh38
rs775424614 Chr.13: 50945431-50945431 on Build GRCh38
rs138649673 Chr.13: 50945479-50945479 on Build GRCh38
rs779468408 Chr.13: 50945531-50945531 on Build GRCh38
rs952624324 Chr.13: 50945394-50945394 on Build GRCh38
rs555463211 Chr.13: 50945487-50945487 on Build GRCh38
rs200390483 Chr.13: 50945438-50945438 on Build GRCh38
rs756300291 Chr.13: 50945544-50945544 on Build GRCh38
rs141367253 Chr.13: 50945480-50945480 on Build GRCh38
rs765368182 Chr.13: 50945478-50945478 on Build GRCh38
rs761310728 Chr.13: 50945407-50945408 on Build GRCh38
rs745471919 Chr.13: 50945505-50945505 on Build GRCh38
rs1042791990 Chr.13: 50945389-50945389 on Build GRCh38
rs201525817 Chr.13: 50945407-50945407 on Build GRCh38
rs952690776 Chr.13: 50945491-50945491 on Build GRCh38
rs370913691 Chr.13: 50945408-50945408 on Build GRCh38
rs368771959 Chr.13: 50945405-50945405 on Build GRCh38
rs756903057 Chr.13: 50945488-50945488 on Build GRCh38
rs77377571 Chr.13: 50945463-50945463 on Build GRCh38
rs566790226 Chr.13: 50945471-50945471 on Build GRCh38
rs944032875 Chr.13: 50945426-50945426 on Build GRCh38
rs769769085 Chr.13: 50945409-50945409 on Build GRCh38
rs759712178 Chr.13: 50945472-50945472 on Build GRCh38
rs1015708068 Chr.13: 50945530-50945530 on Build GRCh38
rs768340427 Chr.13: 50945444-50945444 on Build GRCh38
rs774079225 Chr.13: 50945572-50945572 on Build GRCh38
rs373698054 Chr.13: 50945558-50945558 on Build GRCh38
rs17857128 Chr.13: 50945532-50945532 on Build GRCh38
rs764428875 Chr.13: 50945483-50945483 on Build GRCh38
rs747943029 Chr.13: 50945576-50945576 on Build GRCh38
rs766618424 Chr.13: 50945468-50945468 on Build GRCh38
rs532295786 Chr.13: 50945424-50945424 on Build GRCh38
rs767626668 Chr.13: 50945400-50945400 on Build GRCh38

View Full Product Details