Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112446199-112446472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7386123 c.133G>C p.V45L Chr.12: 112446394-112446394 on Build GRCh38
3456258 c.98C>T p.P33L Chr.12: 112446359-112446359 on Build GRCh38
6947166 c.110A>C p.N37T Chr.12: 112446371-112446371 on Build GRCh38
4718910 c.133G>A p.V45I Chr.12: 112446394-112446394 on Build GRCh38
430369 c.38G>A p.G13D Chr.12: 112446299-112446299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776089364 Chr.12: 112446313-112446313 on Build GRCh38
rs780811696 Chr.12: 112446411-112446411 on Build GRCh38
rs760086740 Chr.12: 112446297-112446297 on Build GRCh38
rs758209360 Chr.12: 112446375-112446375 on Build GRCh38
rs200613531 Chr.12: 112446290-112446290 on Build GRCh38
rs397516808 Chr.12: 112446327-112446327 on Build GRCh38
rs150154839 Chr.12: 112446224-112446224 on Build GRCh38
rs773169250 Chr.12: 112446405-112446405 on Build GRCh38
rs761259292 Chr.12: 112446447-112446447 on Build GRCh38
rs746452065 Chr.12: 112446267-112446267 on Build GRCh38
rs930904396 Chr.12: 112446231-112446231 on Build GRCh38
rs747798804 Chr.12: 112446415-112446415 on Build GRCh38
rs537273886 Chr.12: 112446440-112446440 on Build GRCh38
rs1011511756 Chr.12: 112446241-112446242 on Build GRCh38
rs906966059 Chr.12: 112446362-112446362 on Build GRCh38
rs1006974090 Chr.12: 112446234-112446234 on Build GRCh38
rs768240009 Chr.12: 112446273-112446273 on Build GRCh38
rs779587904 Chr.12: 112446257-112446257 on Build GRCh38
rs756980918 Chr.12: 112446225-112446225 on Build GRCh38
rs1049293041 Chr.12: 112446260-112446260 on Build GRCh38
rs762539984 Chr.12: 112446415-112446416 on Build GRCh38
rs372736227 Chr.12: 112446309-112446309 on Build GRCh38
rs779813529 Chr.12: 112446378-112446378 on Build GRCh38
rs397516795 Chr.12: 112446380-112446380 on Build GRCh38
rs934388335 Chr.12: 112446364-112446364 on Build GRCh38
rs757537175 Chr.12: 112446325-112446325 on Build GRCh38
rs587778635 Chr.12: 112446314-112446314 on Build GRCh38
rs79203122 Chr.12: 112446279-112446279 on Build GRCh38
rs764238610 Chr.12: 112446318-112446318 on Build GRCh38
rs778697402 Chr.12: 112446232-112446232 on Build GRCh38
rs368633510 Chr.12: 112446289-112446289 on Build GRCh38
rs753951666 Chr.12: 112446319-112446319 on Build GRCh38
rs727505378 Chr.12: 112446238-112446238 on Build GRCh38
rs397507500 Chr.12: 112446317-112446317 on Build GRCh38
rs745795958 Chr.12: 112446240-112446240 on Build GRCh38
rs775795422 Chr.12: 112446445-112446445 on Build GRCh38
rs772597897 Chr.12: 112446444-112446444 on Build GRCh38
rs774833026 Chr.12: 112446439-112446439 on Build GRCh38
rs536503257 Chr.12: 112446287-112446287 on Build GRCh38
rs727505387 Chr.12: 112446433-112446433 on Build GRCh38
rs758400048 Chr.12: 112446249-112446249 on Build GRCh38
rs397507502 Chr.12: 112446393-112446393 on Build GRCh38
rs566068139 Chr.12: 112446286-112446286 on Build GRCh38
rs750261927 Chr.12: 112446337-112446337 on Build GRCh38
rs397507501 Chr.12: 112446385-112446385 on Build GRCh38
rs886041585 Chr.12: 112446376-112446376 on Build GRCh38

View Full Product Details