Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75007619-75007890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5927710 c.898G>A p.G300R Chr.14: 75007788-75007788 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023894683 Chr.14: 75007708-75007708 on Build GRCh38
rs113994017 Chr.14: 75007761-75007761 on Build GRCh38
rs775427018 Chr.14: 75007755-75007755 on Build GRCh38
rs751674368 Chr.14: 75007689-75007689 on Build GRCh38
rs900027094 Chr.14: 75007863-75007863 on Build GRCh38
rs751262740 Chr.14: 75007825-75007825 on Build GRCh38
rs892730810 Chr.14: 75007809-75007809 on Build GRCh38
rs766648624 Chr.14: 75007811-75007811 on Build GRCh38
rs780478667 Chr.14: 75007711-75007711 on Build GRCh38
rs536693716 Chr.14: 75007706-75007706 on Build GRCh38
rs764647121 Chr.14: 75007653-75007653 on Build GRCh38
rs752278683 Chr.14: 75007720-75007720 on Build GRCh38
rs376944725 Chr.14: 75007753-75007753 on Build GRCh38
rs773041356 Chr.14: 75007779-75007779 on Build GRCh38
rs756365729 Chr.14: 75007707-75007707 on Build GRCh38
rs746469083 Chr.14: 75007729-75007729 on Build GRCh38
rs770598173 Chr.14: 75007734-75007734 on Build GRCh38
rs764159474 Chr.14: 75007688-75007688 on Build GRCh38
rs762281421 Chr.14: 75007698-75007698 on Build GRCh38
rs142553958 Chr.14: 75007725-75007725 on Build GRCh38
rs773533246 Chr.14: 75007801-75007801 on Build GRCh38
rs767586496 Chr.14: 75007791-75007791 on Build GRCh38
rs781211698 Chr.14: 75007736-75007736 on Build GRCh38
rs761839092 Chr.14: 75007776-75007776 on Build GRCh38
rs774575817 Chr.14: 75007774-75007774 on Build GRCh38
rs763050699 Chr.14: 75007762-75007762 on Build GRCh38
rs763801768 Chr.14: 75007818-75007818 on Build GRCh38
rs745833840 Chr.14: 75007748-75007748 on Build GRCh38
rs758137309 Chr.14: 75007724-75007724 on Build GRCh38
rs768911925 Chr.14: 75007765-75007765 on Build GRCh38
rs553052150 Chr.14: 75007804-75007804 on Build GRCh38
rs984982924 Chr.14: 75007833-75007833 on Build GRCh38
rs756771779 Chr.14: 75007836-75007836 on Build GRCh38
rs763297179 Chr.14: 75007676-75007676 on Build GRCh38
rs369592873 Chr.14: 75007812-75007812 on Build GRCh38
rs1012132094 Chr.14: 75007662-75007662 on Build GRCh38

View Full Product Details