Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037283.1 | NP_001032360.1 | 15 - 16 | 2238 | 99 |
NM_003751.3 | NP_003742.2 | 15 - 16 | 2238 | 99 | |
XM_011515599.1 | XP_011513901.1 | 15 - 16 | 2537 | 99 | |
XM_011515600.1 | XP_011513902.1 | 15 - 16 | 2537 | 99 | |
XM_011515601.1 | XP_011513903.1 | 16 - 17 | 2420 | 99 | |
XM_017012752.1 | XP_016868241.1 | 15 - 16 | 2537 | 99 | |
GenBank mRNA | AK093807.1 | - | 1 - 2 | 1102 | 99 |
AK126670.1 | - | 2 - 3 | 2537 | 99 | |
AK300967.1 | - | 16 - 17 | 1980 | 99 | |
AK302058.1 | - | 16 - 17 | 1764 | 99 | |
BC001173.1 | - | 15 - 16 | 2186 | 99 | |
BC009986.1 | - | 14 - 15 | 1580 | 99 | |
BC110865.1 | - | 15 - 16 | 2164 | 99 | |
U62583.1 | - | 15 - 16 | 2208 | 99 | |
U78525.1 | - | 15 - 16 | 2230 | 99 |