Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037283.1 | NP_001032360.1 | 2 - 3 | 770 | 133 |
NM_003751.3 | NP_003742.2 | 2 - 3 | 770 | 133 | |
XM_011515599.1 | XP_011513901.1 | 2 - 3 | 1069 | 133 | |
XM_011515600.1 | XP_011513902.1 | 2 - 3 | 1069 | 133 | |
XM_011515601.1 | XP_011513903.1 | 3 - 4 | 952 | 133 | |
XM_017012752.1 | XP_016868241.1 | 2 - 3 | 1069 | 133 | |
XM_017012753.1 | XP_016868242.1 | 2 - 3 | 1069 | 133 | |
GenBank mRNA | AK300967.1 | - | 3 - 4 | 512 | 133 |
AK302058.1 | - | 3 - 4 | 296 | 133 | |
BC001173.1 | - | 2 - 3 | 718 | 133 | |
BC009986.1 | - | 1 - 2 | 115 | 133 | |
BC110865.1 | - | 2 - 3 | 696 | 133 | |
BM556863.1 | - | - | 533 | 133 | |
U62583.1 | - | 2 - 3 | 740 | 133 | |
U78525.1 | - | 2 - 3 | 762 | 133 |